Contract Research Organizations To Adopt Ion Torrent Next-Generation Sequencing Platform
MolecularMD and Selah Genomics to Use Oncomine Cancer Research Panel for Clinical Trial Design
Monday, June 2, 2014 11:30 am EDT
Dateline:
CARLSBAD, Calif.
"Content for the Oncomine Cancer Research Panel was selected by leveraging the expertly curated data in the Oncomine Knowledgebase, a long-trusted source of oncology data for pharmaceutical companies, the translational research community, and CROs"
CARLSBAD, Calif.--(BUSINESS WIRE)--Two Contract Research Organizations (CROs) with leading expertise in molecular testing have decided to utilize the new Oncomine Cancer Research panel, which runs on Life Technologies Ion Torrent sequencing instruments, for clinical trial analyses and design.
This comprehensive multi-biomarker assay allows concurrent analyses of DNA and RNA in one workflow, allowing researchers to simultaneously detect all four classes of tumor-driving alterations: single nucleotide variants, copy number variants, tumor suppressors and gene fusions. As CROs and their pharma partners validate their use of this new tool, the potential exists for patients to be more effectively enrolled into clinical trials and study designs made easier.
Founded in 2005 by Brian Druker, M.D., Ph.D. (lead clinical investigator for Gleevec, the first molecularly-targeted anticancer agent) and Sheridan G. Snyder (entrepreneur, founder of Genzyme and Upstate Biotechnology) MolecularMD Corporation develops and commercializes specialty molecular diagnostics for oncology applications across the entire oncology pharmaceutical development life cycle.
Selah Genomics is a clinical diagnostics specialist supporting early stage drug development, clinical trials and regulatory processes in the pharmaceutical industry and helping clinicians and healthcare providers to treat and monitor patients.
“We see the Oncomine Cancer Research Panel as a valuable tool in advancing MolecularMD's comprehensive companion diagnostics program, and personalized medicine in general," stated Glenn Miller, Executive Vice President and CTO of MolecularMD. "The Ion PGM platform, combined with this new panel, meets many of our needs to support drug and diagnostic regulatory submissions with specific actionable genetic markers, while generating a breadth of additional data from variants that can be used in identifying novel drug sensitivities in the future. The Oncomine Cancer Research Panel makes an excellent addition to the broad menu of services we offer to the global biopharmaceutical community.”
“Limited amounts of biopsy tissue is a key challenge in oncology – especially for lung cancer patients,” said Michael Bolick, chief executive officer of Selah Genomics, Inc . “Concurrent analysis of DNA and RNA will reduce the need for repeat biopsy and expand the availability of archived specimens needed for biomarker discovery. We are particularly keen to leverage this technology in development of a universal companion diagnostic for our pharma partners. ”
The Oncomine Cancer Research panel enables simultaneous testing of 160 known cancer genes. The panel employs Life Technologies Ion AmpliSeq chemistry, which requires a very small amount of starting material -- as little as 10 nanograms of DNA or RNA from formalin-fixed paraffin-embedded tissue -- which is often extremely limited in tumor specimens.
“Content for the Oncomine Cancer Research Panel was selected by leveraging the expertly curated data in the Oncomine Knowledgebase, a long-trusted source of oncology data for pharmaceutical companies, the translational research community, and CROs,” said Dan Rhodes, Ph.D., head of oncology strategy, life sciences solutions, for Thermo Fisher. “The combination of one of the world’s largest collection of oncology data with the rapid, simple-to-use Ion sequencing technology positions Thermo Fisher Scientific as a game-changing company in the oncology clinical research space.”
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with revenues of $17 billion and 50,000 employees in 50 countries. Our mission is to enable our customers to make the world healthier, cleaner and safer. We help our customers accelerate life sciences research, solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Through our four premier brands – Thermo Scientific, Life Technologies, Fisher Scientific and Unity Lab Services – we offer an unmatched combination of innovative technologies, purchasing convenience and comprehensive support. For more information, please visit www.thermofisher.com.
Multimedia Files:
Preview image
The Life Technologies Oncomine Cancer Research panels, running on the Ion Torrent Personal Genome Machine, have been adopted by MolecularMD and Selah Genomics. (Photo: Business Wire)
Preview image
Contact:
Thermo Fisher Scientific
Suzanne Clancy, 760-602-4545
suzanne.clancy@thermofisher.com
Thermo Fisher Tweets
Introducing the first NGS-based IVD test for detection of genetic mutations in #NSCLC - Oncomine Dx Target Test… https://t.co/jR0BLLcoT0-3 hours 35 min ago
Hi, Can you plz share serial # of your Vanquish system & your location? We can connect you with the right team. Thanks.-7 hours 25 min ago
Scientists will do just about anything to get an automated cell counter! https://t.co/iWeiF2rvuX See the rebellion. https://t.co/8d3TauNecG-7 hours 57 min ago
What’s the advantage of exceptionally bright readouts for better discrimination of cell subpopulations?… https://t.co/0LbS3hM38m-7 hours 57 min ago
We've just received FDA approval for the first NGS-based IVD test for detection of genetic mutations in #NSCLC https://t.co/uXlmLlmTq1-1 day 1 hour ago