Online Gauge Reduces Radioactive Source Size Requirements for Nuclear Level, Density and Interface Measurements
Thermo Scientific InterfacePRO gauge is designed to improve plant safety and process control 
Wednesday, March 8, 2017 11:33 am EST
Dateline:
FRANKLIN, Mass.
"Employee safety and environmental health are priorities for industrial facility operators who are seeking innovative ways to optimize their plants"
Plant operators in the oil and gas, power, pulp and paper and mining industries can now seek to reduce their on-site radioactive source size requirements by using a nuclear gauge for the density, level and interface measurement of highly viscous process materials.
The Thermo Scientific InterfacePRO gauge can be used for traditional gamma transmission applications (level and density measurements) or in gamma backscattering applications on large process vessels and pipelines. The gauge is designed to improve process control and lower operating costs by enabling quick response to process changes or hazardous conditions. Additionally, the highly-sensitive InterfacePRO gauge uses non-intrusive and durable gamma backscatter technology that is designed to minimize maintenance and lengthen instrument life.
“Employee safety and environmental health are priorities for industrial facility operators who are seeking innovative ways to optimize their plants,” said Lars Andersson, product line manager, level and density gauges, Thermo Fisher Scientific. “By significantly reducing the size of radioactive materials needed on site, the InterfacePRO gauge is designed to help operators improve plant safety and maximize efficiency even in the toughest operating environments.”
The InterfacePRO gauge is a component of the Thermo Scientific NextGEN Nuclear Platform, which also supports the Thermo Scientific DensityPRO and LevelPRO gauges. The InterfacePRO gauge allows users to standardize all process instruments on a single platform with features including:
  • A full suite of communications protocols, including HART, Profibus PA and Foundation Fieldbus, allowing for simplified self-diagnostic, archiving and trend analysis capabilities;
  • Commonality of electronics and spare parts with Thermo Scientific DensityPRO and LevelPRO gauges; and
  • EZCal II software, which offers functions for easy calibration and configuration.
For more information on the Thermo Scientific InterfacePRO gauge, please visit www.thermofisher.com/interfacePRO.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with revenues of $18 billion and more than 55,000 employees globally. Our mission is to enable our customers to make the world healthier, cleaner and safer. We help our customers accelerate life sciences research, solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Through our premier brands – Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific and Unity Lab Services – we offer an unmatched combination of innovative technologies, purchasing convenience and comprehensive support. For more information, please visit www.thermofisher.com.
# # #
Multimedia Files:
Preview image
The Thermo Scientific InterfacePRO gauge is used by plant operators in the oil and gas, power, pulp and paper and mining industries for the density, level and interface measurement of highly viscous process materials.
Contact:
Thermo Fisher Scientific
Marcia Goff, 781 622 1248
Director, Group Communications - Analytical Instruments
marcia.goff@thermofisher.com
Brian Lowe
Greenough
Boston, Mass., USA
+1 617-275-6523
blowe@greenough.biz
Thermo Fisher Tweets
PCR TIPS to obtain the highest sequence accuracy in your PCR applications: https://t.co/meDvZqMKJW https://t.co/roEUTgEIkx-3 hours 59 min ago
The newest software update for the Niton XL5 handheld XRF analyzer includes WiFi. Learn more:… https://t.co/r4LxfsSf04-12 hours 5 min ago
Those look fantastic! Thanks for sharing!-1 day 5 hours ago
DISCOVER the molecular biology research tools that can help advance your science https://t.co/R8k6ZnWTvR https://t.co/7ptqsGxFwF-2 days 8 hours ago
We're really sorry to hear that. Email us at social.media@thermofisher.com.-2 days 10 hours ago