New Energy-Dispersive X-ray Fluorescence Spectrometer Enables Rapid Elemental Analysis in Research and Manufacturing
The Thermo Scientific ARL QUANT’X EDXRF spectrometer is smaller, more sensitive and more user-friendly than previous generations
Category:
Monday, March 6, 2017 6:00 am EST
Dateline:
CHICAGO
"We redesigned the ARL QUANT’X to improve efficiency, ease operation and lower cost of ownership, giving manufacturers and researchers a tool to solve their most challenging analytical tasks."
CHICAGO--(BUSINESS WIRE)--Laboratory managers, quality control technicians and scientists conducting routine analysis or research can now use a new bench-top energy-dispersive x-ray fluorescence (EDXRF) spectrometer to perform faster elemental analysis on a wide range of materials and sample sizes.
The new Thermo Scientific ARL QUANT’X EDXRF spectrometer, which makes its debut at Pittcon 2017 (booth 3021) at McCormick Place, Chicago, is designed to be up to four times more sensitive than the previous version. The instrument covers all periodic table elements, from fluorine to uranium, and provides increased sensitivity and flexibility to determine the full elemental composition of a sample.
The ARL QUANT’X EDXRF spectrometer incorporates a 50W x-ray tube and the latest-generation silicon drift detector (SDD) to enable analysis of light elements and small spot analysis. The instrument requires less lab space than its predecessors and accommodates typical sample sizes used in XRF analysis as well as large and irregularly-shaped samples.
“In materials science and characterization, scientists and quality control technicians are under pressure to perform fast elemental analyses that provide full material identification on many sample types and shapes,” said Didier Bonvin, XRF product manager, Thermo Fisher. “We redesigned the ARL QUANT’X to improve efficiency, ease operation and lower cost of ownership, giving manufacturers and researchers a tool to solve their most challenging analytical tasks.”
The new ARL QUANT’X EDXRF spectrometer provides materials characterization across a variety of applications from petrochemicals, mining and cement to forensics, gemology and environmental controls. Additional benefits of the new ARL QUANT’X EDXRF spectrometer include:
  • WinTrace software on Windows 10, providing a more user-friendly interface that can easily export results to an external computer or LIMS;
  • Standardless analysis through its UniQuant program, which can perform small spot analysis down to 1 mm; and
  • A large sample chamber, allowing the instrument to accept irregularly shaped samples, ranging from 0.5 cm to 40 cm in length and up to 5 cm in height or even bulkier samples of 30 x 40 x 36 cm with the optional sample chamber extension.
For a closer look at the Thermo Scientific ARL QUANT’X EDXRF spectrometer, visit Thermo Fisher at booth 3021, or, for more information, visit www.thermofisher.com/quantx . To learn more about Thermo Fisher’s line of XRF instruments, please visit www.thermofisher.com/XRF.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with revenues of $18 billion and more than 55,000 employees globally. Our mission is to enable our customers to make the world healthier, cleaner and safer. We help our customers accelerate life sciences research, solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Through our premier brands – Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific and Unity Lab Services – we offer an unmatched combination of innovative technologies, purchasing convenience and comprehensive support. For more information, please visit www.thermofisher.com.
Multimedia Files:
Preview image
Thermo Scientific ARL QUANT'X EDXRF (Photo: Business Wire)
Preview image
Contact:
Media Contact Information:
Thermo Fisher Scientific
Nathalie Keng, +41 (0)21 694 72 22
nathalie.keng@thermofisher.com
or
Greenough
Brian Lowe, +1 617-275-6523
blowe@greenough.biz
Thermo Fisher Tweets
Introducing the first NGS-based IVD test for detection of genetic mutations in #NSCLC - Oncomine Dx Target Test… https://t.co/jR0BLLcoT0-3 hours 49 min ago
Hi, Can you plz share serial # of your Vanquish system & your location? We can connect you with the right team. Thanks.-7 hours 39 min ago
Scientists will do just about anything to get an automated cell counter! https://t.co/iWeiF2rvuX See the rebellion. https://t.co/8d3TauNecG-8 hours 11 min ago
What’s the advantage of exceptionally bright readouts for better discrimination of cell subpopulations?… https://t.co/0LbS3hM38m-8 hours 11 min ago
We've just received FDA approval for the first NGS-based IVD test for detection of genetic mutations in #NSCLC https://t.co/uXlmLlmTq1-1 day 1 hour ago