| | |
Thermo Fisher Tweets
Thermo Fisher Scientific Presents XRF Spectrometer for Advanced Materials Characterization
Monday, June 18, 2012 3:41 am EDT
Dateline:
FRANKFURT - ACHEMA 2012
The Thermo Scientific ARL PERFORM’X spectrometer integrates bulk elemental analysis capabilities with mapping and small spot analysis to create a solution that offers unmatched versatility and performance for the analysis of any solid or liquid sample.
The ARL PERFORM’X is a highly precise system offering users a choice of analyzed area size from 35 mm to 0.5 mm. In addition to measuring the standard chemical composition of samples, the system facilitates elemental small spot analysis enabling analysts to pinpoint focus on specimen sections as small as 0.5 mm, significantly improving the versatility of analyses. The instrument’s advanced X-ray mapping and sample visualization capabilities also enable users to identify and characterize inhomogeneities, such as elemental inclusions, defects and gradients in samples previously unidentifiable. The ARL PERFORM’X has the capacity to handle samples of many sizes and types including solids, liquids, powders and thin films, providing laboratories with unrivalled versatility that outperforms other solutions. 
Multimedia Files:
Preview image
Contact:
Aaron Kellogg
1+ (617) 275-6526
akellogg@greenough.biz
Media Contacts
Ron O'Brien
Director, Public Relations
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02454
(781) 622-1242
ron.obrien@thermofisher.com
Tom Keppeler
Public Relations Leader
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02451
(781) 622-1371
thomas.keppeler@thermofisher.com