| | |
Thermo Fisher Tweets
Thermo Fisher Scientific Features Intelligent Process Analyzer at ACHEMA 2012
Category:
Monday, June 18, 2012 2:00 am EDT
Dateline:
FRANKFURT - ACHEMA 2012
"(86.36mm) full-color display. Its bright backlit screen can be seen at nearly any angle and in any lighting condition, making it one of the most versatile in the industry. The interface is clean and easy to understand with intuitive menu -driven design for"
FRANKFURT –ACHEMA 2012– (June 18, 2012) Thermo Fisher Scientific Inc., the world leader in serving science, is featuring its Thermo Scientific AquaPro Multi-Input Intelligent Process Analyzer during ACHEMA 2012. The new AquaPro analyzer features a large full-color display, a plug-and-play architecture, a front panel USB port, up to four channels in one instrument and multiple digital communication options. The AquaPro analyzer is being showcased within Thermo Scientific booth B7, hall 4.2 during ACHEMA 2012, being held from June 18-22 in Frankfurt. 
The AquaPro is a sophisticated and highly flexible analysis platform for a wide range of process applications. Its unique plug-and-play architecture makes it easy to add new features and parameters; all a user needs to do is open the analyzer, slide and click. This simple multi-channel configuration offers the ability to add a simple analog parameter and/or advanced DataStick™ digital sensors to add host communications such as MODBUS, CANopen, Profibus, Ethernet and others. All this comes in one solution and can be done with just a click.
A key feature of the AquaPro analyzer is its large 3.4” (86.36mm) full-color display. Its bright backlit screen can be seen at nearly any angle and in any lighting condition, making it one of the most versatile in the industry. The interface is clean and easy to understand with intuitive menu -driven design for “out of the box” use without the need to study a manual.
The multi-function front panel USB port featured on the AquaPro analyzer offers additional versatility. By plugging in a USB data stick, users can download data from the data logging feature and store it in the AquaPro for easy transfer of settings and parameters to other AquaPro analyzers in the field. The port can also be used to upload new features and updates.
The AquaPro analyzer is programmed with seven user-selectable languages – English, Spanish, French, German, Italian, Portuguese and Chinese. This makes standardizing on the system, anywhere in the world, much easier.  
For more information about the new the Thermo Scientific AquaPro Multi-Input Intelligent Process Analyzer, please visit the Thermo Scientific booth B7 in Hall 4.2 at ACHEMA 2012. Alternatively, please visit www.thermoscientific.com/aquapro. 
For access to all Thermo Fisher news and product photos related to ACHEMA 2012, please visit the online media room at http://news.thermofisher.com
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science. Our mission is to enable our customers to make the world healthier, cleaner and safer. With revenues of $12 billion, we have approximately 39,000 employees and serve customers within pharmaceutical and biotech companies, hospitals and clinical diagnostic labs, universities, research institutions and government agencies, as well as in environmental and process control industries. We create value for our key stakeholders through three premier brands, Thermo Scientific, Fisher Scientific and Unity™ Lab Services, which offer a unique combination of innovative technologies, convenient purchasing options and a single solution for laboratory operations management. Our products and services help our customers solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Visit http://www.thermofisher.com.
# # #
Contact:
James Cairns
+1-978.232.6000
James.cairns@thermofisher.com
Media Contacts
Ron O'Brien
Director, Public Relations
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02454
(781) 622-1242
ron.obrien@thermofisher.com
Tom Keppeler
Public Relations Leader
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02451
(781) 622-1371
thomas.keppeler@thermofisher.com