| | |
Thermo Fisher Tweets
Thermo Fisher Scientific Introduces First Research-Grade FT-IR Spectrometer with One-Touch Operation
Advanced Thermo Scientific Nicolet iS50 FT-IR Platform Integrates Multiple Techniques with Unprecedented Simplicity for Improved Workflows in Analytical Laboratories
Category:
Monday, June 18, 2012 2:00 am EDT
Dateline:
FRANKFURT - ACHEMA 2012
"The powerful OMNIC software suite is capable of converting spectra into composition information without the subjectivity normally associated with spectral interpretation. All of this adds up to an instrument that enables scientists to produce more results in less time than ever before,"
FRANKFURT –ACHEMA 2012– (June 18, 2012) – Thermo Fisher Scientific Inc., the world leader in serving science, is featuring its new Thermo Scientific Nicolet iS50 FT-IR spectrometer, the first research-grade FT-IR with one-touch operation, at ACHEMA 2012. Building on the industry-leading reputation of the Magna, Nexus and Nicolet 6700 spectrometers, the Nicolet iS50 features purpose-built accessories and integrated analytical software capabilities to provide a truly all-in-one materials analysis platform that helps laboratories solve analytical challenges with unprecedented ease. The Nicolet iS50 system is being showcased within Thermo Scientific booth B7, hall 4.2 during ACHEMA 2012, being held from June 18-22 in Frankfurt.
The highly flexible system can be upgraded from a simple FT-IR bench to a fully-automated multi-spectral range system that can acquire spectra from the far- to the near-infrared. Users can initiate novel ATR, Raman and NIR modules at the touch of a button, enabling access to these techniques without manually changing system components.
The research-grade performance of the Nicolet iS50 makes it the ideal choice for academic laboratories. The ease of access to multiple techniques, combined with software designed to take the guesswork out of interpretation, dramatically improves efficiency in analytical laboratories focused on understanding the composition of complex materials.
The flexible system architecture of the Nicolet iS50 is designed to improve the speed of analysis while eliminating the possibility of operational errors that can occur while changing instrument components. The instrument’s integrated diamond ATR allows users to obtain IR spectra in seconds, no matter what other accessories are installed, and a novel in-sample-compartment FT-Raman feature includes a video microstage for point-and-shoot Raman spectroscopy with no fluorescence.
The iS50 also features a new dual-purpose fiber-optic and integrating sphere module that enables collection of NIR spectra from a broad range of bulk samples. When equipped with the automatic beamsplitter exchanger, users can access all three techniques — ATR, Raman and NIR — by pressing the unique one-touch buttons. This simplified operation makes the instrument ideal for laboratories worldwide, allowing scientists working in diverse fields, from pharmaceutical formulation to polymer development, forensics and art conservation, to answer more questions in less time and with greater confidence.  
“Today’s scientists must be more productive and possess a broader skillset than ever before,” said Simon Nunn, global marketing director for Molecular Spectroscopy at Thermo Fisher. “In its simplest form, the Nicolet iS50 is a high-performance yet simple-to-use FT-IR spectrometer. When equipped with its optional modules, it becomes a powerful materials analysis workbench capable of extracting an incredible amount of information from a wide range of samples. All techniques are available at the scientist’s fingertips — literally.”
The Nicolet iS50 analyzes complex mixtures and materials using its GC-IR or TGA-IR capabilities. The new OMNIC Mercury software automatically isolates and identifies compounds eluted from a GC column or evolved from a TGA. This functionality is especially useful in TGA, where several compounds can evolve at the same time. Such analyses provide insight into subtle formulation differences in plastics and rubbers, which can lead to not-so-subtle differences in product performance and quality.
“The powerful OMNIC software suite is capable of converting spectra into composition information without the subjectivity normally associated with spectral interpretation. All of this adds up to an instrument that enables scientists to produce more results in less time than ever before,” added Nunn.
The Nicolet iS50R, a full-featured research variant of the Nicolet iS50, is designed for advanced applications including vibrational circular dichroism, PM-IRRAS and time-resolved spectroscopy. 
The Nicolet iS50 comes standard with automated performance verification using NIST-traceable standards, enabling high-quality and consistent data every day.
For more information about the new Thermo Scientific Nicolet iS50 FT-IR spectrometer, please visit Thermo Scientific booth B7 in Hall 4.2 at ACHEMA 2012. Alternatively, please call +1 866-463-6522, email analyze@thermofisher.com or visit www.thermoscientific.com/iS50. 
For access to all Thermo Fisher news and product photos related to ACHEMA 2012, please visit the online media room at http://news.thermofisher.com.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. (NYSE: TMO) is the world leader in serving science. Our mission is to enable our customers to make the world healthier, cleaner and safer. With revenues of $12 billion, we have approximately 39,000 employees and serve customers within pharmaceutical and biotech companies, hospitals and clinical diagnostic labs, universities, research institutions and government agencies, as well as in environmental and process control industries. We create value for our key stakeholders through three premier brands, Thermo Scientific, Fisher Scientific and Unity™ Lab Services, which offer a unique combination of innovative technologies, convenient purchasing options and a single solution for laboratory operations management. Our products and services help our customers solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Visit http://www.thermofisher.com.
###
Contact:
Media Contact:
Aaron Kellogg
+1-617-275-6526
akellogg@greenough.biz
Customer Contact:
Amy Knock
+1-608- 276-6114
amy.knock@thermofisher.com
Media Contacts
Ron O'Brien
Director, Public Relations
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02454
(781) 622-1242
ron.obrien@thermofisher.com
Tom Keppeler
Public Relations Leader
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02451
(781) 622-1371
thomas.keppeler@thermofisher.com