| | |
Thermo Fisher Tweets
Thermo Fisher Scientific Launches Smaller, More Efficient Twin-Screw Extruder
Thermo Scientific Process 11 Has Reduced Footprint, Uses Small Quantities of Sample Material and Saves Cost, Time and Labor in Polymer and Food Industry Research
Category:
Monday, June 18, 2012 2:00 am EDT
Dateline:
FRANKFURT - ACHEMA 2012
"To our customers, laboratory space is at a premium and cost control is critical, so we’ve designed the Process 11 extruder to address both issues,"
FRANKFURT –ACHEMA 2012– (June 18, 2012) – Thermo Fisher Scientific Inc., the world leader in serving science, is featuring its new Thermo Scientific Process 11 parallel co-rotating twin-screw extruder for polymer and food industry research applications at ACHEMA 2012. The 11 mm extruder is designed to minimize material costs, be easier to use and optimize laboratory space. To achieve this, the benchtop Process 11 extruder uses a minimal amount of sample material (20 g) and features a user-friendly touchscreen with integrated feeder control. The Process 11 extruder is being showcased within Thermo Scientific booth B7, hall 4.2 during ACHEMA 2012, being held from June 18-22 in Frankfurt.
“To our customers, laboratory space is at a premium and cost control is critical, so we’ve designed the Process 11 extruder to address both issues,” said Karl Gerhard Hoppmann, vice president and general manager of Thermo Fisher’s material characterization business. “Our compact, standalone design is perfect for small-scale experiments yet robust enough to deliver results that are relevant to production conditions. The results are easily obtained due to the full scalability of the Thermo Scientific standalone compounding systems.” 
The new Process 11 features a throughput of 20 g/h to 2.5 kg/h and a segmented screw design with removable top half barrel. The machine’s compact monocoque design makes its footprint four to five times smaller than competing twin-screw extruders, and it enables easy transport and fume hood applications. The screw elements and barrel design scale geometrically across the whole suite of Thermo Scientific extruders and allow easy process scale-up. The instrument is available immediately.
Thermo Fisher, one of the pioneers in rheology, successfully supports a wide range of industries with its comprehensive Thermo Scientific material characterization solutions. Material characterization solutions analyze and measure viscosity, elasticity, processability and temperature-related mechanical changes of plastics, food, cosmetics, pharmaceuticals and coatings, chemical or petrochemical products, plus a wide variety of liquids or solids. For more information, please visit www.thermoscientific.com/mc
For more information about the new Thermo Scientific Process 11 parallel co-rotating twin-screw extruder, please visit Thermo Scientific booth B7 in Hall 4.2 at ACHEMA 2012. Alternatively, please call +1 866-463-6522, email analyze@thermofisher.com or visit www.thermoscientific.com/mc.
For access to all Thermo Fisher news and product photos related to ACHEMA 2012, please visit the online media room at http://news.thermofisher.com.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science. Our mission is to enable our customers to make the world healthier, cleaner and safer. With revenues of $12 billion, we have approximately 39,000 employees and serve customers within pharmaceutical and biotech companies, hospitals and clinical diagnostic labs, universities, research institutions and government agencies, as well as in environmental and process control industries. We create value for our key stakeholders through three premier brands, Thermo Scientific, Fisher Scientific and Unity™ Lab Services, which offer a unique combination of innovative technologies, convenient purchasing options and a single solution for laboratory operations management. Our products and services help our customers solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Visit www.thermofisher.com.
# # # 
Contact:
Petra Roth
Thermo Fisher Scientific
Dieselstr. 4, 76227 Karlsruhe/Germany
+49 (0) 721 40 94 169
petra.roth@thermofisher.com
www.thermoscientific.com/mc
Secondary Contact Information:
Aaron Kellogg
+1-617- 275-6526
akellogg@greenoughcom.com
Media Contacts
Ron O'Brien
Director, Public Relations
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02454
(781) 622-1242
ron.obrien@thermofisher.com
Tom Keppeler
Public Relations Leader
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02451
(781) 622-1371
thomas.keppeler@thermofisher.com