Thermo Fisher Scientific Releases Next-Generation Benchtop NMR Spectrometer
Thermo Scientific picoSpin 80 NMR Spectrometer Breaks New Ground in Benchtop Analysis at 80 MHz
Tuesday, August 27, 2013 9:00 am EDT
Dateline:
MADISON, Wis.
"The picoSpin 80 provides nearly double the analytical power of the picoSpin 45, producing even more detailed spectra, marking another step in our continuing commitment to make high-quality NMR spectrometers available to as many scientists as possible."
MADISON, Wis.--(BUSINESS WIRE)--Academic and industrial laboratories can now easily collect routine high-resolution nuclear magnetic resonance (NMR) data on molecules in solution using the Thermo Scientific picoSpin 80, a portable, competitively-priced NMR spectrometer built for benchtop operation. The instrument’s magnet, rated at 2 Tesla, is designed to provide superior resolution of chemical and structural characteristics not available using lower magnetic field strengths.
Based on published specifications, the Thermo Scientific picoSpin 80 spectrometer features the highest resolution of many widely-known, currently available benchtop NMR instruments, offering scientists a complement to the already popular Thermo Scientific picoSpin 45 spectrometer.
“The picoSpin 80 and picoSpin 45 are game-changing instruments that make NMR analysis possible for more scientists, especially in academic, industrial or R&D laboratories,” said Simon Nunn, global marketing director, molecular spectroscopy, Thermo Fisher Scientific. “The picoSpin 80 provides nearly double the analytical power of the picoSpin 45, producing even more detailed spectra, marking another step in our continuing commitment to make high-quality NMR spectrometers available to as many scientists as possible.”
The picoSpin 80 spectrometer features and benefits include:
  • Temperature-controlled magnet that does not require liquid cryogens;
  • Lightweight, portable design intended to make it easy to share the instrument across multiple laboratories;
  • Easy-to-operate fluid capillary system that does not require NMR tubes or other consumables.
Thermo Scientific is one of the premier brands of Thermo Fisher Scientific Inc. More information about the Thermo Scientific picoSpin 80 can be found at www.picospin.com.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science. Our mission is to enable our customers to make the world healthier, cleaner and safer. With revenues of $13 billion, we have 39,000 employees and serve customers within pharmaceutical and biotech companies, hospitals and clinical diagnostic labs, universities, research institutions and government agencies, as well as in environmental and process control industries. We create value for our key stakeholders through three premier brands, Thermo Scientific, Fisher Scientific and Unity Lab Services, which offer a unique combination of innovative technologies, convenient purchasing options and a single solution for laboratory operations management. Our products and services help our customers solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Visit www.thermofisher.com.
Multimedia Files:
Preview image
Thermo Scientific picoSpin 80 is a portable, competitively-priced NMR spectrometer built for benchtop operation. (Photo: Business Wire)
Preview image
The picoSpin 80 and picoSpin 45 are game-changing instruments that make NMR analysis possible for more scientists. (Photo: Business Wire)
Contact:
For Thermo Fisher Scientific Inc.
Media:
Amy Knock, +1-608-276-6114
Amy.Knock@thermofisher.com
or
Aaron Kellogg, +1-617-275-6526
akellogg@greenough.biz
Thermo Fisher Tweets
Did you miss us at #AGBT17? We presented data on 600 bp reads with the Ion S5 System - check it out here! https://t.co/5YYW2FSfez-1 day 9 hours ago
New to cloning, PCR, or reverse transcription? Free online learning resources to help get you started… https://t.co/LD0zZisZNG-2 days 20 hours ago
The time has come for life science researchers to consider themselves as biobankers https://t.co/0rpMwl1FcY https://t.co/22cHtLLDja-3 days 11 min ago
Want GENTLE cell detachment? LEARM MORE more about TrypLE reagents: https://t.co/otKw8Ab1N0 https://t.co/SCIjnEhmaF-3 days 3 hours ago
PCR TIP: Simplify your PCR reaction setup with hot-start PCR. Ditch the ice bucket. https://t.co/cLDVEKE8Si https://t.co/ZnGA3YZVjQ-4 days 2 hours ago