| | |
Thermo Fisher Tweets
Thermo Fisher Scientific Showcases Benchtop Chemistry Analyzer at AACC 2012
Category:
Tuesday, July 17, 2012 2:47 pm EDT
Dateline:
LOS ANGELES, Calif.
LOS ANGELES, Calif. (July 17, 2012) -- Thermo Fisher Scientific Inc., the world leader in serving science, is showcasing the Thermo Scientific Indiko, a fully automated, compact, random access chemistry analyzer at the 2012 annual conference of the American Association for Clinical Chemistry (AACC). Indiko™ is an easy to use, cost-effective system designed for clinical chemistry and specialty testing and is being featured within Thermo Scientific booth #2215 during AACC, being held at the Los Angeles Convention Center from July 17-19.
The Thermo Scientific Indiko benchtop analyzer is designed for routine clinical chemistry testing in small laboratory settings and specialty testing such as specific proteins, drug of abuse testing and therapeutic drug monitoring including immunosuppressant drug monitoring (ISD). Bar-coded, ready to use Thermo Scientific system reagents provide ease-of-use and optimized package sizes for low volume testing needs. A mix of bar-coded primary tubes and sample cups may be used at any time, increasing the flexibility of operation. Patient oriented testing produces results quickly thus enhancing the quality of patient care.
Indiko includes several user friendly automated features, like advanced dilution management and real-time monitoring of reagent usage, which help to manage a daily workload fluently. Samples, reagents and consumables can continuously be loaded without interrupting the testing process, and different sample types can be analyzed at the same time. A real-time QC program with multiple Westgard Rules assures the reliable performance. Application data as well as calibrator and control values are loadable from a file or from a two-dimensional barcode by the barcode scanner. The test flow definition allows for up to four reagent additions in each test, offering automation even for the most complex methods. The unique cuvette design supports low reagent volumes thus reducing operating costs, especially important when working with specialty tests. Each cuvette is used only once to ensure accurate and precise results.
Indiko has a small footprint and fits ideally in a laboratory with space limitations. The intuitive user interface has a highly graphical design and is supported by a touch screen. Besides routine clinical chemistry testing, Thermo Scientific Indiko serves various dedicated specialty testing needs by offering quality, flexibility and automation and thus saving time and money.
For more information about Thermo Scientific Idiko analyzer, please visit Thermo Scientific booth #2215 at AACC 2012. Alternatively, call (510) 979-5000, email sales.diagnostics.fmt@thermofisher.com or visit http://www.thermoscientific.com/indiko.
For access to all Thermo Fisher Scientific news and product photos related to AACC 2012, please visit the online media room at http://www.thermofisher.com/AACC2011.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. (NYSE: TMO) is the world leader in serving science. Our mission is to enable our customers to make the world healthier, cleaner and safer. With revenues of $12 billion, we have approximately 39,000 employees and serve customers within pharmaceutical and biotech companies, hospitals and clinical diagnostic labs, universities, research institutions and government agencies, as well as in environmental and process control industries. We create value for our key stakeholders through three premier brands, Thermo Scientific, Fisher Scientific and Unity™ Lab Services, which offer a unique combination of innovative technologies, convenient purchasing options and a single solution for laboratory operations management. Our products and services help our customers solve complex analytical challenges, improve patient diagnostics and increase laboratory productivity. Visit www.thermofisher.com.
# # #
Contact:
Kathy Ruzich, Manager Marcom
510-979-5157
kathy.ruzich@thermofisher.com
www.thermoscientific.com/Diagnostics
Media Contacts
Ron O'Brien
Director, Public Relations
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02454
(781) 622-1242
ron.obrien@thermofisher.com
Tom Keppeler
Public Relations Leader
Thermo Fisher Scientific
81 Wyman Street
Waltham, MA 02451
(781) 622-1371
thomas.keppeler@thermofisher.com